Merck
Mifamurtide: osteosarcoma: ineffective and harmful.
Prescrire international (2011-06-09)
ABSTRACT
The standard treatment for children and young adults with osteosarcoma consists of surgery, preceded and followed by methotrexate-based chemotherapy. Mifamurtide is an immunostimulant derived from a bacterial cell wall component. It is authorised in the European Union as an adjunct to combination chemotherapy after complete excision of non-metastatic osteosarcoma. Only one comparative, unblinded trial has been published, and its design was particularly complex. In a study population of 678 patients, adding mifamurtide to chemotherapy after tumour excision did not prolong the overall 6-year survival rate, which was about 75% with both treatments. Only serious adverse effects were collected, and they were not systematically recorded. Hypersensitivity reactions occurred in clinical trials, along with pleural and pericardial effusions, seizures, and muscle spasms. Severe hearing loss occurred in 12% of the patients treated with mifamurtide in the comparative trial, versus 7% of the other patients. In practice, given the lack of any survival benefit and the risk of serious adverse effects, it is better not to add mifamurtide to chemotherapy regimens used for treatment of osteosarcoma.