Merck
  • Promoted negative staining of proteins in SDS-PAGE using Eosin B compounded with magnesium chloride.
Promoted negative staining of proteins in SDS-PAGE using Eosin B compounded with magnesium chloride.
Electrophoresis (2010-11-17)
Xu Wang, Sun-Young Hwang, Wei-Tao Cong, Li-Tai Jin, Jung-Kap Choi
ABSTRACT
In this study, we describe an effective visualizing technique for proteins in SDS-PAGE based on the organic dye, Eosin B, the sensitivity of which can be further strengthened by the addition of magnesium to the staining solution after electrophoresis. The newly developed protocol is low in cost and easily performed compared with the common methods for protein analysis in 1-D and 2-D gels. It provides a much better sensitivity (0.2 ng of single protein band) than that of imidazole-zinc negative stain for fixing and staining within 1 h, and an excellent performance in terms of compatibility with MALDI-TOF MS. The results show that similar identification scores and numbers of matched peptides were obtained by both methods. Furthermore, the effects of different metal salts on the quality of protein visualization by Eosin B were also investigated. Because of its sensitivity, stability, and safety, this stain may be a more practical method for protein determination in the routine laboratory.
Sigma-Aldrich
Eosin B, certified by the Biological Stain Commission, Dye content 90 %