Merck
  • Novel amidino substituted 2-phenylbenzothiazoles: synthesis, antitumor evaluation in vitro and acute toxicity testing in vivo.
Novel amidino substituted 2-phenylbenzothiazoles: synthesis, antitumor evaluation in vitro and acute toxicity testing in vivo.
Bioorganic & medicinal chemistry (2010-01-12)
Livio Racané, Marijeta Kralj, Lidija Suman, Ranko Stojković, Vesna Tralić-Kulenović, Grace Karminski-Zamola
ABSTRACT
The efficient synthesis of new bis-substituted nitro-amidino, amino-amidino (10a, 10b-13a, 13b) and previously prepared diamidino 2-phenyl-benzothiazoles (9a, 9b) is described. The compounds 11a and 11b were prepared by recently developed methodology of the key precursors in zwitterionic form 8a and 8b with 4-nitrobenzoylchloride in a very good yield (70%). All compounds except diamidino-substituted 2-phenylbenzothiazole 9a show exceptionally prominent tumor cell-growth inhibitory activity and cytotoxicity, whereby the special selectivity of amino-amidine 2-phenylbenzothiazole 12a towards MCF-7 and H 460 cells makes this compound a prospective lead compound that should be further evaluated in animal models. All in vivo tested compounds (12a, 12b, 13a and 13b) are absorbed from mice gastrointestinal system. LD(50) are between 67.33 and 696.2mg/kg body weight (OECD/EPA toxicity categories 2-3).
Sigma-Aldrich
2-Phenylbenzothiazole, 97%