Merck
  • The use of oral contraceptives as a prevention of recurrent premenstrual psychosis.
The use of oral contraceptives as a prevention of recurrent premenstrual psychosis.
Psychiatry research (2009-10-20)
Marta Coromina Sadurni, Judith Usall Rodie, Laia Miquel de Montagut, Mònica Sánchez Autet
ABSTRACT
Premenstrual psychosis is a rare and not formally recognized disorder (DSM-IVR, ICD-10). The literature mainly consists of clinical cases. There have been preliminary reports of improvement in such cases after administration of oral contraceptives. We present a case of premenstrual psychosis in which hormonal treatment was effective in preventing symptomatic relapses.
Sigma-Aldrich
Ethynodiol diacetate