Merck
  • Analysis of vasodilator responses to peroxynitrite in the hindlimb vascular bed of the cat.
Analysis of vasodilator responses to peroxynitrite in the hindlimb vascular bed of the cat.
Journal of cardiovascular pharmacology (2007-12-01)
Bobby D Nossaman, Trinity J Bivalacqua, Hunter C Champion, Syed R Baber, Philip J Kadowitz
ABSTRACT
The free radical peroxynitrite (ONOO-) is formed in biological systems from the reaction of nitric oxide (NO) with superoxide (O2-) and can react with protein and nonprotein thiol groups to produce tissue injury. However, these pathologic actions of (ONOO-) may have been overemphasized, in that (ONOO-) has vasorelaxant properties through activation of soluble guanylate cyclase; inhibits leukocyte-endothelial cell interactions; and reduces ischemia-reperfusion injury in the heart, lung, and liver. It has been reported that tolerance develops to the vasodilator actions of (ONOO-) and that (ONOO-) impairs vascular function. However, little, if anything, is known about responses to (ONOO-) in the hindlimb circulation of the cat. To better understand the effects of (ONOO-) on responses to vasoactive agonists and the mechanism by which (ONOO-) induces vasodilation, the effects of short-term exposure to (ONOO-) were investigated under constant-flow conditions in the hindlimb vascular bed of the cat. In these studies, direct intraarterial injections of (ONOO-) produced dose-dependent decreases in hindquarters perfusion pressure. The vasodilator responses to (ONOO-) were rapid in onset, were short in duration, and could be repeated without exhibiting tachyphylaxis. Vasodilator responses to (ONOO-) were not changed in the presence of inhibitors of nitric-oxide synthase, cyclooxygenase, or K+-ATP (adenosine triphosphate-sensitive potassium) channels. Furthermore, responses to (ONOO-) were enhanced in duration by the type 5-cGMP (cyclic guanosine monophosphate) phosphodiesterase inhibitor zaprinast, whereas rolipram, a type 4-cGMP phosphodiesterase inhibitor, was without effect. Repeated administration of (ONOO-) had no significant effect on responses to vasoconstrictor or to vasodilator agents including acetylcholine. These results show that (ONOO-) has significant vasodilator activity in the hindlimb vascular bed of the cat and suggest that the response is mediated by a cGMP- dependent mechanism. The results of experiments with repeated injections of (ONOO-) indicate that (ONOO-) does not impair vasoconstrictor and endothelium-dependent or endothelium-independent vasodilator responses. Furthermore, tolerance did not develop with repeated short-term exposure to (ONOO-). Moreover, the results of experiments with inhibitors suggest that responses to (ONOO-) are not dependent on K-ATP (adenosine triphosphate-sensitive potassium) channel activation, increased NOS activity, or the formation of products in the cyclooxygenase pathway. The results of these studies are consistent with the hypothesis that (ONOO-) is rapidly converted in the hindlimb circulation to a substance that has the properties of an NO donor. These studies suggest that under physiologic conditions, the cytotoxic effects of (ONOO-) on a short-term basis may have been overemphasized.