- Competitive photoinduced electron transfer by the complex formation of porphyrin with cyclodextrin bearing viologen.
Competitive photoinduced electron transfer by the complex formation of porphyrin with cyclodextrin bearing viologen.
Chemical communications (Cambridge, England) (2006-10-13)
Wei Deng, Takeshi Onji, Hiroyasu Yamaguchi, Noriaki Ikeda, Akira Harada
PMID17031435
ABSTRACT
Photoinduced electron transfer between a porphyrin and a new guest cyclodextrin bearing viologen occurs by a supramolecular formation with conformational change of a guest molecule.
Sigma-Aldrich
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), Dye content 75 %