Merck
  • The influence of spacer-containing enzyme conjugate on the sensitivity and specificity of enzyme immunoassays for hapten.
The influence of spacer-containing enzyme conjugate on the sensitivity and specificity of enzyme immunoassays for hapten.
Clinica chimica acta; international journal of clinical chemistry (2005-12-13)
Anupam Basu, Seema Nara, Shail K Chaube, Kiran Rangari, Kiran P Kariya, Tulsidas G Shrivastav
ABSTRACT
In hapten enzyme immunoassays (EIA), there is an increase or decrease of labeled hapten recognition by antibody that affects sensitivity of the assay. We incorporated a spacer between a hapten derivative and enzyme to test its influence on the sensitivity and specificity of enzyme immunoassays. Antibodies were generated against cortisol-3-O-carboxymethyl-oxime-bovine serum albumin (cortisol-3-O-CMO-BSA) and cortisol-21-hemisuccinate-bovine serum albumin (cortisol-21-HS-BSA) as an immunogen. Four cortisol horseradish peroxidase (HRP) enzyme conjugates were prepared using 2 cortisol derivatives (cortisol-3-O-CMO and cortisol-21-HS) with and without adipic acid dihydrazide (ADH) as a spacer. Eight combinations of homologous and heterologous assays were evaluated. The incorporation of ADH spacer in cortisol-enzyme conjugate improved the sensitivity in heterologous (bridge and site plus bridge) EIA systems. In heterologous assays (site plus bridge), the presence of spacer in enzyme conjugate reduced the cross-reactivity with cross-reacting steroids. Spacer in the enzyme conjugate for hapten ELISA can improve the sensitivity of heterologous assay of hapten-like steroids. It may also reduce the cross-reactivity for some assays.
Sigma-Aldrich
Adipic acid dihydrazide, ≥98% (titration)