Merck
  • Probabilistic model for estimating field mortality of target and non-target bird populations when simultaneously exposed to avicide bait.
Probabilistic model for estimating field mortality of target and non-target bird populations when simultaneously exposed to avicide bait.
Pest management science (2005-03-05)
John J Johnston, Melvin J Holmes, Andy Hart, Dennis J Kohler, Randal S Stahl
ABSTRACT
A probabilistic model was developed to estimate target and non-target avian mortality associated with the application of the avicide CPTH (3-chloro-p-toluidine hydrochloride) to minimize sprouting rice damage in the southern USA. CPTH exposures for individual birds were predicted by random sampling from species-specific non-parametric distributions of bait seed consumption and CPTH residues detected on individual bait seeds. Mortality was predicted from the species-specific exposure versus mortality relationship. Individual variations in this response were captured in the model by Monte Carlo sampling from species-specific distributions of slopes and median toxicity values (LD50) for each bird. The model was used to simultaneously predict mortality (percentage of exposed population and number of birds killed/weight of consumed bait) for a target (blackbird) and non-target (mourning dove) species feeding on bait sites for up to five consecutive days.
Sigma-Aldrich
2-Chloro-4-aminotoluene, 98%