Merck
  • Biosynthesis of the angiogenesis inhibitor borrelidin by Streptomyces parvulus Tü4055: cluster analysis and assignment of functions.
Biosynthesis of the angiogenesis inhibitor borrelidin by Streptomyces parvulus Tü4055: cluster analysis and assignment of functions.
Chemistry & biology (2004-04-29)
Carlos Olano, Barrie Wilkinson, César Sánchez, Steven J Moss, Rose Sheridan, Vidya Math, Alison J Weston, Alfredo F Braña, Christine J Martin, Markiyan Oliynyk, Carmen Méndez, Peter F Leadlay, José A Salas
ABSTRACT
The biosynthetic gene cluster for the angiogenesis inhibitor borrelidin has been cloned from Streptomyces parvulus Tü4055. Sequence analysis indicates that the macrolide ring of borrelidin is formed by a modular polyketide synthase (PKS) (borA1-A6), a result that was confirmed by disruption of borA3. The borrelidin PKS is striking because only seven rather than the nine modules expected for a nonaketide product are encoded by borA1-A6. The starter unit of the PKS has been verified as trans-cyclopentane-1,2-dicarboxylic acid (trans-1,2-CPDA), and the genes involved in its biosynthesis identified. Other genes responsible for biosynthesis of the nitrile moiety, regulation, and self-resistance were also identified.
Sigma-Aldrich
Borrelidin, from Streptomyces parvulus, ≥98% (HPLC)