Merck
  • [Infrared and Raman spectra studies on isomultinuclear organometallic carbyne compounds].
[Infrared and Raman spectra studies on isomultinuclear organometallic carbyne compounds].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-30)
Z Xu, A Mayr
ABSTRACT
The infrared and Raman spectra of five isomultinuclear organometallic carbyne compounds were studied. The results show that the introduction of N atom from TMEDA or P atom from DPPE to W atom (both N and P are strong sigma-donor and weak pi-acceptor) will decrease the bonding intensity of W identical to C. The pressure tunning IR and Raman spectra of compound III indicate the presence of pressure-induced phase-transition near 20 kbar in it. There are two effects on the nu (W identical to C) of this compound with increasing pressure: the normal increasing the nu (W identical to C) wavenumber due to the shorting W identical to C bond and increasing the W-CO pi backbonding to decrease the nu (W identical to C) wavenumber.