Merck
  • Effects of commercial antiglaucoma drugs to glutamate-induced [Ca2+)]i increase in cultured neuroblastoma cells.
Effects of commercial antiglaucoma drugs to glutamate-induced [Ca2+)]i increase in cultured neuroblastoma cells.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2003-06-28)
Show-Jen Hong, Kwou-Yeung Wu, Hwei-Zu Wang, Jim C Fong
ABSTRACT
Over releasing of glutamate and cellular calcium influx always results in neuronal death. In the present study, we investigated various commercial antiglaucoma drugs including timolol (0.58 microM to 58 microM), betaxolol (1.62 microM to 162 microM), carteolol (6.8 microM to 680 microM), pilocarpine (4.08 microM to 408 microM), latanoprost (0.01 microM to 1.1 microM), dorzolamide (6.16 microM to 616 microM), brinzolamide (2.6 microM to 260 microM), brimonidine (0.68 microM to 68 microM), dipivefrin (0.28 microM to 28 microM) and preservative benzalkonium chloride on their effects to inhibit glutamate-induced intracellular free Ca(2+) ([Ca(2+)](i)) increase in cultured N1E-115 neuroblastoma cells. These drugs were diluted from original concentrations to 1/100, 1/1000 and 1/10000. The [Ca(2+)](i) mobility was studied after loading with fura-2-AM and analyzed by spectrofluorometry. It was found that betaxolol, dipivefrin and brimonidine have remarkable effects not only to inhibit the glutamate-induced [Ca(2+)](i) increase but also to decrease the basal [Ca(2+)](i). In the case of other drugs, only high concentration of timolol (58 microM) exhibited significant effect to completely prevent glutamate-induced [Ca(2+)](i) increase. Moreover, benzalkonium chloride did not exhibit any inhibitive effect. These results indicate that betaxolol, dipivefrin and brimonidine may have neuroprotective effects to inhibit the glutamate-induced over Ca(2+) influx damage.
Product Description
Dipivefrine for system suitability, European Pharmacopoeia (EP) Reference Standard
Dipivefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard