- Antimicrobial compounds of low molecular mass are constitutively present in insects: characterisation of beta-alanyl-tyrosine.
Antimicrobial compounds of low molecular mass are constitutively present in insects: characterisation of beta-alanyl-tyrosine.
The number of bacterial and fungal strains that have developed resistance against the classical antibiotics continues to grow. The intensified search for new antibiotic lead compounds has resulted in the discovery of numerous endogenous peptides with antimicrobial properties in plants, bacteria and animals. Their possible applications as anti-infective agents are often limited by their size, in reference to production costs and susceptibility to proteases. In this article, we report recent isolations of antimicrobial compounds from insects, with molecular masses less than 1 kDa. Experimental approaches are discussed and the first data on the antimicrobial properties of beta-alanyl-tyrosine (252 Da), one of such low molecular mass compounds isolated from the fleshfly Neobellieria bullata, are presented. We also offer evidence for the constitutive presence of antimicrobial compounds in insects of different orders, in addition to the previously identified inducible antimicrobial peptides.