Merck
  • Modeling of deoxy- and dideoxyaldohexopyranosyl ring puckering with MM3(92).
Modeling of deoxy- and dideoxyaldohexopyranosyl ring puckering with MM3(92).
Carbohydrate research (2001-10-12)
W M Rockey, M K Dowd, P J Reilly, A D French
ABSTRACT
Extensive variations of the ring structures of three deoxyaldohexopyranoses, L-fucose, D-quinovose, and L-rhamnose, and four dideoxyaldohexopyranoses, D-digitoxose, abequose, paratose, and tyvelose, were studied by energy minimization with the molecular mechanics algorithm MM3(92). Chair conformers, 4C(1) in D-quinovose and the equivalent 1C(4) in L-fucose and L-rhamnose, overwhelmingly dominate in the three deoxyhexoses; in the D-dideoxyhexoses, 4C(1) is again dominant, but with increased amounts of 1C(4) forms in the alpha anomers of the three 3,6-dideoxyhexoses, abequose, paratose, and tyvelose and in both alpha and beta anomers of the 2,6-dideoxyhexose D-digitoxose. In general, modeled proton-proton coupling constants agreed well with experimental values. Computed anomeric ratios strongly favor the beta configuration except for D-digitoxose, which is almost equally divided between alpha and beta configurations, and L-rhamnose, where the beta configuration is somewhat favored. MM3(92) appears to overstate the prevalence of the equatorial beta anomer in all three deoxyhexoses, as earlier found with fully oxygenated aldohexopyranoses.