Merck
  • The durability of adhesion to phosphoric acid etched, wet dentin substrates.
The durability of adhesion to phosphoric acid etched, wet dentin substrates.
Dental materials : official publication of the Academy of Dental Materials (1999-06-24)
G Kato, N Nakabayashi
ABSTRACT
The purpose of this study was to investigate the influence of remaining non-resin-impregnated, phosphoric acid demineralized dentin upon the long-term durability of specimens that were wet-bonded to bovine dentin substrates. Prepared bovine dentin samples were etched with 65% phosphoric acid then rinsed with water and kept wet during application of 5 wt% 4-methacryloyloxyethyl trimellitate anhydride (4-META) in acetone primer. This was followed by application of a photocured dentin-bonding agent consisting of 4-methacryloyloxyethyl trimellitate anhydride/triethyleneglycol dimethacrylate-camphorquinone/N-phenylglycine (4-META/TEGDMA-CQ/NPG). The tensile bond strength (TBS) of bonded specimens was determined after immersion in 37 degrees C water for various time intervals. Generated data were analyzed for statistical significance by one-way ANOVA and Duncan's New Multiple Range Test (p < 0.05). The dentin side of the tensile-load-fractured specimens was examined under optical and scanning electron microscopes (SEM). TBS decreased from 6.6 +/- 1.0 MPa after 1-day water immersion to 3.4 +/- 1.7 MPa after 1 month of water immersion. After 6 months of water immersion, TBS was found to be 3.9 +/- 0.9 MPa and this decreased to 2.0 +/- 1.0 MPa for specimens immersed in water for 1 year, a statistically significant difference (p < 0.05). Optical microscopic and SEM observations disclosed failure patterns within demineralized, non-resin-impregnated dentin that increased with the period of water immersion. The bond durability to wet dentin was poor when demineralized dentin was not resin-impregnated, resulting in exposure of collagen fibrils which hydrolyzed during long periods of water immersion.
Sigma-Aldrich
N-Phenylglycine, 97%