Merck
  • Germicidal activity of a chlorous acid-chlorine dioxide teat dip and a sodium chlorite teat dip during experimental challenge with Staphylococcus aureus and Streptococcus agalactiae.
Germicidal activity of a chlorous acid-chlorine dioxide teat dip and a sodium chlorite teat dip during experimental challenge with Staphylococcus aureus and Streptococcus agalactiae.
Journal of dairy science (1998-09-28)
R L Boddie, S C Nickerson, R W Adkinson
ABSTRACT
Three postmilking teat dips were tested for efficacy against Staphylococcus aureus and Streptococcus agalactiae in two separate studies using experimental challenge procedures that were recommended by the National Mastitis Council. The first study evaluated a barrier teat dip product containing chlorous acid-chlorine dioxide as the germicidal agent, and the second study evaluated a sodium chlorite product with a barrier component as well as a sodium chlorite product without a barrier component. The chlorous acid-chlorine dioxide teat dip reduced new intramammary infections (IMI) caused by Staph. aureus by 91.5% and reduced new IMI caused by Strep. agalactiae by 71.7%. The barrier dip containing sodium chlorite reduced new IMI caused by Staph. aureus and Strep. agalactiae by 41.0 and 0%, respectively. The nonbarrier dip containing sodium chlorite reduced new IMI caused by Staph. aureus by 65.6% and reduced new IMI caused by Strep. agalactiae by 39.1%. Teat skin and teat end conditions were evaluated before and after the second study; no deleterious effects among dipped quarters compared with control quarters were noted for the two sodium chlorite products.
Sigma-Aldrich
Sodium chlorite, puriss. p.a., 80% (RT)