Merck
  • Microbial hexachlorobenzene dechlorination under three reducing conditions.
Microbial hexachlorobenzene dechlorination under three reducing conditions.
Chemosphere (1998-09-24)
B V Chang, C J Su, S Y Yuan
ABSTRACT
The potential dechlorination of hexachlorobenzene (HCB) in medium by 1,2,3-trichlorobenzene (TCB)-adapted mixed culture under three reducing conditions was investigated. It was found that strongest to weakest HCB dechlorination occurred in the order of methanogenic conditions > sulfate-reducing conditions > denitrifying conditions. Under denitrifying conditions, no dechlorination was observed during the first 20 days of incubation. Biotransformation occurred in this order: HCB-->pentachlorobenzene (PCB)-->1,2,3,5-tetrachlorobenzene (TeCB)-->1,3,5-TCB + 1,2,4-TCB-->1,3-dichlorobenzene (DCB), HCB dechlorination was delayed following treatment with ferric chloride and manganese dioxide, but enhanced by the addition of lactate and pyruvate under methanogenic or sulfate-reducing conditions, the addition of acetate had no significant effect on HCB dechlorination under any of the three reducing conditions. Sequential dechlorination was observed at concentrations of 2-50 mg/L, but at a significantly slower rate at the highest concentrations.
Sigma-Aldrich
1,3-Dichlorobenzene, 98%
Sigma-Aldrich
1,3-Dichlorobenzene, puriss., ≥99.0% (GC)
Supelco
1,2,3,5-Tetrachlorobenzene, PESTANAL®, analytical standard