Merck
  • Ligand-induced conformational changes modify proteolytic cleavage of the adipocyte insulin-sensitive glucose transporter.
Ligand-induced conformational changes modify proteolytic cleavage of the adipocyte insulin-sensitive glucose transporter.
The Biochemical journal (1993-10-01)
Y Yano, J M May
ABSTRACT
The transport conformation of the human erythrocyte glucose transporter (GLUT1) modifies rates of proteolytic cleavage of this protein by a variety of enzymes. We investigated the effects of ligand-induced conformational change on the susceptibility to enzymic cleavage of the insulin-sensitive rat adipocyte glucose transporter (GLUT4). A GLUT4-enriched slow sedimenting microsomal fraction was prepared from basal adipocytes and subjected to PAGE and immunoblotting. The GLUT4 protein was detected in these immunoblots with a C-terminal-specific antiserum as an M(r)-46,000-50,000 doublet. GLUT1 protein was not detected by a GLUT1-specific antiserum in these membranes. Tryptic digestion caused loss of the GLUT4 signal in immunoblots in a time- and concentration-dependent fashion. Low-M(r) membrane-bound fragments were not observed in electrophoretic gels, whether detection was attempted by immunoblotting or by counting radioactivity in gel slices following photolabelling with [3H]cytochalasin B. Transport-specific ligands known to induce an outward-facing conformation in the human erythrocyte GLUT1 protein retarded cleavage of the GLUT4 protein by submaximal concentrations of trypsin, whereas ligands known to induce an inward-facing conformation increased the extent of cleavage. The transported substrate D-glucose retarded tryptic cleavage of GLUT4. This result contrasts with the known behaviour of GLUT1, in which D-glucose accelerates cleavage. Cleavage of GLUT4 by thermolysin was also retarded by the outward-binding analogue 4,6-O-ethylidene glucose. These results show that the conformational sensitivity to proteolysis of GLUT4 mirrors that of GLUT1, except that the glucose-loaded GLUT4 has a different steady-state configuration, which may reflect underlying kinetic differences between the two proteins.
Sigma-Aldrich
4,6-O-Ethylidene-α-D-glucose