Merck
  • Evaluation of new medium with chromogenic substrates for members of the family Enterobacteriaceae in urine samples.
Evaluation of new medium with chromogenic substrates for members of the family Enterobacteriaceae in urine samples.
Journal of clinical microbiology (1995-01-01)
H Kodaka, M Ishikawa, M Iwata, F Kashitani, S Mizuochi, K Yamaguchi
ABSTRACT
A new medium containing 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt (Glu agar) for Escherichia coli and a new medium containing 5-bromo-3-indolyl-beta-D-galactoside (Gal agar) for beta-galactosidase-positive members of the family Enterobacteriaceae were compared with MacConkey agar in a diagnostic trial with 3,562 urine specimens. The isolation rates of E. coli and beta-galactosidase-positive Enterobacteriaceae were increased 8.4 and 19.5%, respectively. The sensitivities and specificities of Glu agar and Gal agar were 98.5 and 100% and 99.2 and 99.5%, respectively.
Sigma-Aldrich
Bluo-Gal, reagent for selection of recombinant bacterial clones
Sigma-Aldrich
5-Bromo-3-indolyl β-D-galactopyranoside, ≥98%