Merck
  • Synthesis of novel 1H-1,2,3-triazole tethered C-5 substituted uracil-isatin conjugates and their cytotoxic evaluation.
Synthesis of novel 1H-1,2,3-triazole tethered C-5 substituted uracil-isatin conjugates and their cytotoxic evaluation.
European journal of medicinal chemistry (2012-11-06)
Kewal Kumar, Sunil Sagar, Luke Esau, Mandeep Kaur, Vipan Kumar
ABSTRACT
The present manuscript describes the synthesis of uracil-isatin hybrids via azide-alkyne cycloadditions and their cytotoxic evaluation against three human cancer cell lines viz. HeLa (cervix), MCF-7 (breast) and DU145 (prostate) using MTT assay. The evaluation studies revealed the dependence of cytotoxicity on C-5 substituents of both uracil and isatin as well as the alkyl chain length with compounds 6g and 6k showing IC(50) values 18.21 and 13.90 μM respectively against DU145 cell lines. Most of the synthesized conjugates exhibited considerable selectivity against MCF-7 and DU145 cell lines.
Sigma-Aldrich
Isatin, 97%