Merck
  • 19F molecular MR imaging for detection of brain tumor angiogenesis: in vivo validation using targeted PFOB nanoparticles.
19F molecular MR imaging for detection of brain tumor angiogenesis: in vivo validation using targeted PFOB nanoparticles.
Angiogenesis (2012-10-12)
Céline Giraudeau, Françoise Geffroy, Sébastien Mériaux, Fawzi Boumezbeur, Philippe Robert, Marc Port, Caroline Robic, Denis Le Bihan, Franck Lethimonnier, Julien Valette
ABSTRACT
Molecular imaging with magnetic resonance imaging (MRI) targeted contrast agents has emerged as a promising diagnostic approach in cancer research to detect associated biomarkers. In this work, the potential of (19)F MRI was investigated to detect angiogenesis with α(ν)β(3)-targeted perfluorooctylbromide nanoparticles (PFOB NP) in a U87 glioblastoma mouse model at 7 Tesla. Mice were injected intravenously with targeted or non-targeted NP and (19)F images were immediately acquired for 90 min using a PFOB-dedicated MRI sequence. Mice infused with targeted NP exhibited higher concentrations in tumors than mice of the control group, despite the presence of nonspecific signal originating from the blood. Imaging results were corroborated by histology and fluorescence imaging, suggesting specific binding of targeted NP to α(ν)β(3) integrin. Two other groups of mice were injected 24 h before imaging to allow blood clearance but no significant differences were found between both groups, probably due to a loss of specificity of PFOB NP. This is the first demonstration of the ability of (19)F MRI to detect α(ν)β(3)-integrin endothelial expression in brain tumors in vivo.
Sigma-Aldrich
1-Bromoheptadecafluorooctane, 99%