Merck
  • Excess electron transfer dynamics in DNA hairpins conjugated with N,N-dimethylaminopyrene as a photosensitizing electron donor.
Excess electron transfer dynamics in DNA hairpins conjugated with N,N-dimethylaminopyrene as a photosensitizing electron donor.
Chemical communications (Cambridge, England) (2012-10-06)
Man Jae Park, Mamoru Fujitsuka, Haruhiro Nishitera, Kiyohiko Kawai, Tetsuro Majima
ABSTRACT
Excess electron transfer dynamics in DNA hairpins was investigated by femtosecond laser flash photolysis of a donor-DNA-acceptor system using N,N-dimethylaminopyrene and diphenylacetylene as an electron donor and acceptor, respectively. It was revealed that the excess electron hopping rate between T's is faster than that of the hole.
Sigma-Aldrich
1-Aminopyrene, 97%