MilliporeSigma
  • [Effect and mechanism of curcumol on angiogenesis activity of zebrafishes].
[Effect and mechanism of curcumol on angiogenesis activity of zebrafishes].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-09-25)
Lili Tian, Jianyong Dong, Changjiang Huang
ABSTRACT
To conduct a preliminary study on the effect of curcumol in promoting angiogenesis activity and its mechanism in zebrafishes, in order to provide basis for clinical prescription. Zebrafishes biological model was established to, observe curcumol's effect on embryo blood vessel growth, blood vessel regeneration of adult fishes after tail-cutting and tissue regeneration of fish fries after tail-cutting. The relative fluorescence quantitative PCR method was adopted to determine the gene expression of vascular endothelial growth factor (VEGFA) and receptor VEGFR2 of fish fries after tail-cutting. Curcumol contributed to angiogenesis of intersegmental blood vessels in zebrafishes embryos and speed up regeneration of blood vessels in adult fishes after tail-cutting. Furthermore, curcumol can increase the gene expression of VEGFA and VEGFR2 in fish fries. Curcumol can promote angiogenesis in zebrafishes, and enhance the gene expression of VEGFA and VEGFR2 in fish fries after tail-cutting and speed up the regeneration of their tails.