Merck
  • Preconcentration of metal ions through chelation on a synthesized resin containing O, O donor atoms for quantitative analysis of environmental and biological samples.
Preconcentration of metal ions through chelation on a synthesized resin containing O, O donor atoms for quantitative analysis of environmental and biological samples.
Environmental monitoring and assessment (2012-07-20)
Aminul Islam, Mohammad Asaduddin Laskar, Akil Ahmad
ABSTRACT
Nascent Amberlite XAD-4 has been used as the polymeric support for the synthesis of a stable extractor of metal ions, by incorporating phthalic acid through azo bridging. Elemental analyses and infra-red spectral and thermal studies were carried out for its characterization. The water regain value and hydrogen ion capacity were found to be 12.50 and 5.75 mmol g(-1), respectively. The optimum pH range for the maximum sorption of Ni(II), Mn(II), Cu(II), Zn(II), Cd(II), Cr(III), and Co(II) was observed at pH 5.5-8.0 with the corresponding half-loading time (t (1/2)) of 9, 5, 9, 9, 3, 9, and 5 min, respectively. The preconcentration factor for Ni(II), Mn(II), Cu(II), Zn(II), Cd(II), Cr(III), and Co(II) are 190, 190, 190, 180, 180, 160, and 160, with the corresponding limit of preconcentration in the range of 5.25-6.25 μg L(-1). The detection limits, for flame atomic absorption spectrophotometry, were found to be 0.62, 0.60, 0.65, 0.75, 0.72, 0.84, and 0.85 μg L(-1), respectively. Method has been successfully applied to the analysis of water samples, multivitamin formulations, infant food substitutes, hydrogenated oil, and fishes.
Product Description
Supelco
Amberlite XAD4
Supelco
Amberlite XAD4, 20-60 mesh