Merck
  • NGF promotes long-term memory formation by activating poly(ADP-ribose)polymerase-1.
NGF promotes long-term memory formation by activating poly(ADP-ribose)polymerase-1.
Neuropharmacology (2012-07-10)
Shao-Hui Wang, Xiao-Mei Liao, Dan Liu, Juan Hu, Yang-Yang Yin, Jian-Zhi Wang, Ling-Qiang Zhu
ABSTRACT
Nerve growth factor (NGF) is a critical secreted protein that plays an important role in development, survival, and function of the mammalian nervous system. Previously reports suggest that endogenous NGF is essential for the hippocampal plasticity/memory and NGF deprivation induces the impairment of hippocampus-related memory and synaptic plasticity. However, whether exogenous supplement of NGF could promote the hippocampus-dependent synaptic plasticity/memory and the possible underlying mechanisms are not clear. In this study we found that NGF administration facilitates the hippocampus-dependent long-term memory and synaptic plasticity by increasing the activity of PARP-1, a polymerase mediating the PolyADP-ribosylation and important for the memory formation. Co-application of 3-Aminobenzamide (3-AB), a specific inhibitor of PARP-1, distinctly blocked the boosting effect of NGF on memory and synaptic plasticity, and the activation of downstream PKA-CREB signal pathway. Our data provide the first evidence that NGF supplement facilitates synaptic plasticity and the memory ability through PARP-1-mediated protein polyADP-ribosylation and activation of PKA-CREB pathway.
Sigma-Aldrich
3-Aminobenzamide, ≥99% (TLC)