Merck
  • Lentivirus capture directly from cell culture with Q-functionalised microcapillary film chromatography.
Lentivirus capture directly from cell culture with Q-functionalised microcapillary film chromatography.
Journal of chromatography. A (2012-07-10)
N J Darton, D Darling, M J Townsend, D J McNally, F Farzaneh, N K H Slater
ABSTRACT
A new disposable adsorbent material for fast anion-exchange capture of nano-complexes without prefiltering, clarification or pre-processing of samples was developed based on plastic microcapillary films (MCFs). An MCF containing 19 parallel microcapillaries, each with a mean internal diameter of 142 ± 10 μm, was prepared using a melt extrusion process from an ethylene-vinyl alcohol copolymer (EVOH). The MCF internal surfaces were functionalised using branched chain chemistries to attach quaternary amine groups producing an anion-exchange adsorbent. The purification of nano-complexes using this newly fabricated MCF-EVOH-Q was successfully demonstrated with the capture of lentivirus from pre-filtered culture harvest. This 5m chromatographic substrate was found to bind and elute ∼40% of bound lentivirus or 2.5 × 10(6)infectious units (ifu). The unique properties of this chromatographic substrate that allow the passage of large particulates was further demonstrated with the capture of lentiviral particles from unfiltered un-processed culture media containing cells and cell debris. Using this approach, 56% or 1 × 10(7)ifu of captured lentivirus was eluted. A device based on this new material might be used at an early stage in clinical lentiviral production to harvest lentiviral particles, directly from bioreactors.
Sigma-Aldrich
Poly(vinyl alcohol-co-ethylene), ethylene 32 mol %