Merck
  • Quaternized magnetic microspheres for the efficient removal of reactive dyes.
Quaternized magnetic microspheres for the efficient removal of reactive dyes.
Water research (2012-06-26)
Chendong Shuang, Penghui Li, Aimin Li, Qing Zhou, Mancheng Zhang, Yang Zhou
ABSTRACT
In this paper, a novel quaternized magnetic resin, NDMP, was prepared and characterized. Two reactive dyes (RDs), Orange G (OG) and red RWO, were used as a small-molecular RD and large-molecular RD, respectively, to investigate their adsorption on NDMP. A common quaternized magnetic resin, MIEX, was selected for comparison. The adsorption kinetics of OG onto both resins and the adsorption kinetics of RWO onto NDMP followed pseudo-second-order kinetics, whereas the adsorption of RWO onto MIEX was better fitted by pseudo-first-order kinetics. The experimental data illustrated that the equilibrium adsorption amount of both RDs onto NDMP (1.9 mmol OG/g, 0.70 mmol RWO/g) was twice as large as that on MIEX (1.0 mmol OG/g, 0.35 mmol RWO/g). The Langmuir equation and the Freundlich model fit the isotherm data for OG and RWO adsorption, respectively. The adsorption of OG on the NDMP and MIEX resins declined in the presence of NaCl or Na₂SO₄. The effects of the salts on the adsorption of RWO were different. The recyclability of NDMP and MIEX were also evaluated. This work provides a reusable efficient adsorbent for the removal of RDs.
Sigma-Aldrich
Orange G, certified by the Biological Stain Commission, Dye content 80 %
Sigma-Aldrich
Orange G, certified by the Biological Stain Commission
Sigma-Aldrich
Orange G, for NA electrophoresis