Merck
Off-label uses of trazodone: a review.
Expert opinion on pharmacotherapy (2012-06-21)
Letizia Bossini, Ilaria Casolaro, Despoina Koukouna, Federica Cecchini, Andrea Fagiolini
ABSTRACT
Trazodone is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors. It is approved by the FDA for the treatment of depression. Insomnia is the most frequent reason for prescription of trazodone. It has also been proven useful in the treatment of anxiety disorders. Other off-label uses include the treatment of bulimia, benzodiazepine/alcohol dependence, fibromyalgia, central nervous system degenerative diseases (behavioral disorders in dementia and other organic disorders), schizophrenia, chronic pain disease and diabetic neuropathy, sexual dysfunction. This paper evaluates trazodone's efficacy and safety in its off-label uses. It also discusses the possibility that a combination of trazodone with SSRIs may prevent or treat some of the SSRI side effects, such as anxiety, insomnia and sexual dysfunction, in addition to synergically increasing SSRIs' antidepressant activity. Few clinical trials have been conducted to evaluate trazodone's efficacy in the treatment of the diseases and symptoms for which it is often used in clinical practice. More studies are necessary to investigate possible new therapeutic indications, and to scientifically demonstrate the risk/benefit ratio for the many conditions for which trazodone is used, but not approved by the FDA.
Sigma-Aldrich
Trazodone hydrochloride, ≥99% (HPLC), powder
Supelco
Trazodone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®