Merck
  • Purification and characterization of ZmRIP1, a novel reductant-inhibited protein tyrosine phosphatase from maize.
Purification and characterization of ZmRIP1, a novel reductant-inhibited protein tyrosine phosphatase from maize.
Plant physiology (2012-04-25)
Bingbing Li, Yanxia Zhao, Liyan Liang, Huibo Ren, Yu Xing, Lin Chen, Mingzhu Sun, Yuanhua Wang, Yu Han, Haifeng Jia, Conglin Huang, Zhongyi Wu, Wensuo Jia
ABSTRACT
Protein tyrosine phosphatases (PTPases) have long been thought to be activated by reductants and deactivated by oxidants, owing to the presence of a crucial sulfhydryl group in their catalytic centers. In this article, we report the purification and characterization of Reductant-Inhibited PTPase1 (ZmRIP1) from maize (Zea mays) coleoptiles, and show that this PTPase has a unique mode of redox regulation and signaling. Surprisingly, ZmRIP1 was found to be deactivated by a reductant. A cysteine (Cys) residue (Cys-181) near the active center was found to regulate this unique mode of redox regulation, as mutation of Cys-181 to arginine-181 allowed ZmRIP1 to be activated by a reductant. In response to oxidant treatment, ZmRIP1 was translocated from the chloroplast to the nucleus. Expression of ZmRIP1 in Arabidopsis (Arabidopsis thaliana) plants and maize protoplasts altered the expression of genes encoding enzymes involved in antioxidant catabolism, such as At1g02950, which encodes a glutathione transferase. Thus, the novel PTPase identified in this study is predicted to function in redox signaling in maize.
Sigma-Aldrich
Phenylarsine oxide, ≥97%, powder