Merck
  • Lewis acid-catalyzed intramolecular [3+2] cycloaddition of cyclopropane 1,1-diesters with alkynes for the synthesis of cyclopenta[c]chromene skeletons.
Lewis acid-catalyzed intramolecular [3+2] cycloaddition of cyclopropane 1,1-diesters with alkynes for the synthesis of cyclopenta[c]chromene skeletons.
Chemistry, an Asian journal (2012-04-11)
Xiao-Feng Xia, Xian-Rong Song, Xue-Yuan Liu, Yong-Min Liang
ABSTRACT
An efficient method to construct cyclopenta[c]chromene skeletons by Lewis acid-catalyzed intramolecular [3+2] cycloaddition of cyclopropane 1,1-diesters with alkynes is presented. Two new fused cycles can be formed in one step in moderate to excellent yields (up to 94 %), and the products can be converted into bioactive barbituric acid derivatives (1) under simple reaction conditions.
Sigma-Aldrich
Barbituric acid, ReagentPlus®, 99%
Supelco
Barbituric acid, for spectrophotometric det. of cyanide, ≥99.5%
Sigma-Aldrich
Sodium barbiturate, ≥97.0% (T)