Merck
  • Revised recommendations for the measurements of the serum iron in human blood. Iron Panel of the International Committee for Standardization in Haematology.
Revised recommendations for the measurements of the serum iron in human blood. Iron Panel of the International Committee for Standardization in Haematology.
British journal of haematology (1990-08-01)
ABSTRACT
Certain modifications to the previously published ICSH recommendations (1978) have been approved by the Iron Panel of the International Committee for Standardization in Haematology (ICSH). They include substitution of the chromogen bathophenanthroline sulphonate with ferrozine or ferene, which are more sensitive and cheaper, and a reduction in the volume of the test sample from 2 ml to 0.5 ml.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, Suitable for determination of Fe
Supelco
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, for spectrophotometric det. of Fe, ≥99.0%