Merck
  • Antitumor effect of liposomal histone deacetylase inhibitor-lipid conjugates in vitro.
Antitumor effect of liposomal histone deacetylase inhibitor-lipid conjugates in vitro.
Chemical & pharmaceutical bulletin (2011-11-02)
Yoshiyuki Hattori, Yasuo Nagaoka, Manami Kubo, Haruka Yamasaku, Yuta Ishii, Hiroko Okita, Hiroki Nakano, Shinichi Uesato, Yoshie Maitani
ABSTRACT
Histone deacetylase inhibitor (HDACI), suberoylanilide hydroxamic acid (SAHA), approved by the Food and Drug Administration (FDA) for the treatment of cutaneous T cell lymphoma, is a promising new treatment strategy for various cancers. In this study, we hypothesized that a liposomal formulation of HDACI might efficiently deliver HDACI into tumors. To incorporate HDACI efficiently into the liposomal membrane, we synthesized six HDACI-lipid conjugates, in which polyethylene glycol(2000) (PEG(2000))-lipid or cholesterol (Chol) was linked with a potent hydroxamic acid, HDACI, SAHA or K-182, by cleavable linkers, such as ester, carbamide and disulfide bonds. Liposomal HDACI-lipid conjugates were prepared with distearoylphosphatidylcholine (DSPC) and HDACI-Chol conjugate or with DSPC, Chol and HDACI-PEG-lipid conjugates, and their cytotoxicities were evaluated for human cervix tumor HeLa and mouse colon tumor Colon 26 cells. Among the liposomes, liposomal oleyl-PEG(2000)-SAHA conjugated with SAHA and oleyl-PEG(2000) via a carbamate linker showed higher cytotoxicity via hyperacetylation of histone H3 and induction of caspase 3/7 activity. These results suggested that liposomal HDACI-lipid conjugates may be a potential tool for cancer therapy.
Sigma-Aldrich
DL-α-Phosphatidylcholine, distearoyl, ≥98%
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphocholine, ≥99%