- Expanding the repertoire of natural product-inspired ring pairs for molecular recognition of DNA.
Expanding the repertoire of natural product-inspired ring pairs for molecular recognition of DNA.
Organic letters (2011-10-01)
Katy A Muzikar, Jordan L Meier, Daniel A Gubler, Jevgenij A Raskatov, Peter B Dervan
PMID21957930
ABSTRACT
A furan amino acid, inspired by the recently discovered proximicin natural products, was incorporated into the scaffold of a DNA-binding hairpin polyamide. While unpaired oligomers of 2,4-disubstituted furan amino acids show poor DNA-binding activity, furan (Fn) carboxamides paired with N-methylpyrrole (Py) and N-methylimidazole (Im) rings demonstrate excellent stabilization of duplex DNA as well as discrimination of noncognate sequences, consistent with function as a Py mimic according to the Py/Im polyamide pairing rules.
Sigma-Aldrich
Netropsin dihydrochloride, from Streptomyces netropsis, ≥98% (HPLC and TLC), powder