Merck
[Studies on the alkaloids of Senecio scandens growing in Guangdong].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-10-01)
Xiao-Fang Guo, Meng-Hua Liu, Wei Peng, Yong-Gang Wang, Cui-Ping Yang, Wei-Wei Su
ABSTRACT
To study alkaloids of Senecio scandens growing in Guangdong. The rapid resolution liquid chromatography-electrospray ionization mass spectrometry (RRLC-ESI-MS/MS) was used to analyse alkaloids of Senecio scandens growing in Guangdong, and senkirkine was isolated and purified by silica gel column chromatography. Four alkaloids were identified as senkirkine, dehydrosenkirkine, monocrotaline and adonifoline, and senkirkine was firstly isolated from Senecio scandens growing in Guangdong. Senkirkine is the main component of Senecio scandens growing in Guangdong.