Merck
  • Self-organization of 1-methylnaphthalene on the surface of artificial snow grains: a combined experimental-computational approach.
Self-organization of 1-methylnaphthalene on the surface of artificial snow grains: a combined experimental-computational approach.
The journal of physical chemistry. A (2011-09-03)
Dominik Heger, Dana Nachtigallová, František Surman, Ján Krausko, Beata Magyarová, Miroslav Brumovský, Miroslav Rubeš, Ivan Gladich, Petr Klán
ABSTRACT
A combined experimental-computational approach was used to study the self-organization and microenvironment of 1-methylnaphthalene (1MN) deposited on the surface of artificial snow grains from vapors at 238 K. The specific surface area of this snow (1.1 × 10(4) cm(2) g(-1)), produced by spraying very fine droplets of pure water from a nebulizer into liquid nitrogen, was determined using valerophenone photochemistry to estimate the surface coverage by 1MN. Fluorescence spectroscopy at 77 K, in combination with molecular dynamics simulations, and density functional theory (DFT) and second-order coupled cluster (CC2) calculations, provided evidence for the occurrence of ground- and excited-state complexes (excimers) and other associates of 1MN on the snow grains' surface. Only weak excimer fluorescence was observed for a loading of 5 × 10(-6) mol kg(-1), which is ∼2-3 orders of magnitude below monolayer coverage. However, the results indicate that the formation of excimers is favored at higher surface loadings (>5 × 10(-5) mol kg(-1)), albeit still being below monolayer coverage. The calculations of excited states of monomer and associated moieties suggested that a parallel-displaced arrangement is responsible for the excimer emission observed experimentally, although some other associations, such as T-shape dimer structures, which do not provide excimer emission, can still be relatively abundant at this surface concentration. The hydrophobic 1MN molecules, deposited on the ice surface, which is covered by a relatively flexible quasi-liquid layer at 238 K, are then assumed to be capable of dynamic motion resulting in the formation of energetically preferred associations to some extent. The environmental implications of organic compounds' deposition on snow grains and ice are discussed.
Sigma-Aldrich
1-Methylnaphthalene, ≥95%
Sigma-Aldrich
1-Methylnaphthalene, 95%
Supelco
1-Methylnaphthalene, analytical standard