Merck
  • Investigation of the effect of a variety of pulse errors on spin I=1 quadrupolar alignment echo spectroscopy.
Investigation of the effect of a variety of pulse errors on spin I=1 quadrupolar alignment echo spectroscopy.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-06-15)
Xiang Ma, Cheng Sun, Gregory S Boutis
ABSTRACT
We report on an analysis of a well known three-pulse sequence for generating and detecting spin I=1 quadrupolar order when various pulse errors are taken into account. In the situation of a single quadrupolar frequency, such as the case found in a single crystal, we studied the potential leakage of single and/or double quantum coherence when a pulse flip error, finite pulse width effect, RF transient or a resonance offset is present. Our analysis demonstrates that the four-step phase cycling scheme studied is robust in suppressing unwanted double and single quantum coherence as well as Zeeman order that arise from the experimental artifacts, allowing for an unbiased measurement of the quadrupolar alignment relaxation time, T(1Q). This work also reports on distortions in quadrupolar alignment echo spectra in the presence of experimental artifacts in the situation of a powdered sample, by simulation. Using our simulation tool, it is demonstrated that the spectral distortions associated with the pulse artifacts may be minimized, to some extent, by optimally choosing the time between the first two pulses. We highlight experimental results acquired on perdeuterated hexamethylbenzene and polyethylene that demonstrate the efficacy of the phase cycling scheme for suppressing unwanted quantum coherence when measuring T(1Q). It is suggested that one employ two separate pulse sequences when measuring T(1Q) to properly analyze the short time behavior of quadrupolar alignment relaxation data.