Merck
  • Cross-link formation between mutant galectins of Caenorhabditis elegans with a substituted cysteine residue and asialofetuin via a photoactivatable bifunctional reagent.
Cross-link formation between mutant galectins of Caenorhabditis elegans with a substituted cysteine residue and asialofetuin via a photoactivatable bifunctional reagent.
Biological & pharmaceutical bulletin (2011-06-02)
Mayumi Tamura, Tomoharu Takeuchi, Takamasa Nonaka, Ken-Ichi Kasai, Yoichiro Arata
ABSTRACT
LEC-1 is the first tandem repeat-type galectin isolated from an animal system; this galectin has two carbohydrate recognition domains in a single polypeptide chain. Because its two lectin domains have different sugar-binding profiles, these domains are thought to interact with different carbohydrate ligands. In our previous study, we showed that a mutant of LEC-1 in which a cysteine residue was introduced at a unique position in the N-terminal lectin domain (Nh) can be cross-linked with a model glycoprotein ligand, bovine asialofetuin, by using a bifunctional photoactivatable cross-linking reagent, benzophenone-4-maleimide. In the present work, we applied the same procedure to the C-terminal lectin domain (Ch) of LEC-1. Cross-linked products were formed in the cases of two mutants in which a cysteine residue was introduced at Lys¹⁷⁷ and Ser²⁶⁸, respectively. This method is very useful for capturing and assigning endogenous ligand glycoconjugates with relatively low affinities to each carbohydrate recognition domain of the whole tandem repeat-type galectin molecule.