Merck
  • Design of a metal primer containing a dithiooctanoate monomer and a phosphonic acid monomer for bonding of prosthetic light-curing resin composite to gold, dental precious and non-precious metal alloys.
Design of a metal primer containing a dithiooctanoate monomer and a phosphonic acid monomer for bonding of prosthetic light-curing resin composite to gold, dental precious and non-precious metal alloys.
Dental materials journal (2011-05-21)
Kunio Ikemura, Toshihide Fujii, Noriyuki Negoro, Takeshi Endo, Yoshinori Kadoma
ABSTRACT
The effect of metal primers on adhesion of a resin composite to dental metal alloys was investigated. Experimental primers containing a dithiooctanoate monomer [10-methacryloyloxydecyl 6,8-dithiooctanoate (10-MDDT) or 6-methacryloyloxyhexyl 6,8-dithiooctanoate (6-MHDT)] and a phosphonic acid monomer [6-methacryloyloxyhexyl phosphonoacetate (6-MHPA) or 6-methacryloyloxyhexyl 3-phosphonopropionate (6-MHPP)] were prepared. After treating Au, Au alloy, Ag alloy, Au-Ag-Pd alloy, and Ni-Cr alloy with the experimental primers, their shear bond strengths (SBSs) with a prosthetic light-curing resin composite (Solidex, Shofu Inc., Japan) were measured after 1-day storage followed by 5,000 thermal cycles. The SBSs between Solidex and the primer-treated metals which were incubated in air at 50°C for 2 months were further measured. Results showed that the SBSs [mean (SD)] of all metal adherends treated with primer DT-PA-1 (5.0 wt% 10-MDDT, 1.0 wt% 6-MHPA) ranged between 31.2 (5.2) and 34.5 (5.8) MPa. The SBSs of the primer-treated metals did not degrade after 2-month incubation at 50°C. Therefore, a combined primer application consisting of a dithiooctanoate monomer and a phosphonic acid monomer provided efficacious bonding to Au as well as precious and non-precious metal alloys.
Sigma-Aldrich
Phosphonoacetic acid, 98%
Sigma-Aldrich
3-Phosphonopropionic acid, technical grade, 94%