Merck
  • A cyclometalated iridium(III) complex with enhanced phosphorescence emission in the solid state (EPESS): synthesis, characterization and its application in bioimaging.
A cyclometalated iridium(III) complex with enhanced phosphorescence emission in the solid state (EPESS): synthesis, characterization and its application in bioimaging.
Dalton transactions (Cambridge, England : 2003) (2011-02-02)
Huazhou Wu, Tianshe Yang, Qiang Zhao, Jing Zhou, Chunyan Li, Fuyou Li
ABSTRACT
Iridium(III) complexes with intense phosphorescence in solution have been widely applied in organic light-emitting diodes, chemosensors and bioimaging. However, little attention has been paid to iridium(III) complexes showing weak phosphorescence in solution and enhanced phosphorescence emission in the solid state (EPESS). In the present study, two β-diketonate ligands with different degrees of conjugation, 1-phenyl-3-methyl-4-benzoyl-5-pyrazolone (HL1) and 1-phenyl-3-methyl-4-phenylacetyl-5-pyrazolone (HL2), have been synthesized to be used as ancillary ligands for two iridium(III) complexes, Ir(ppy)(2)(L1) and Ir(ppy)(2)(L2) (Hppy = 2-phenylpyridine). The two complexes have been characterized by single-crystal X-ray crystallography, (1)H NMR and elemental analysis. Interestingly, Ir(ppy)(2)(L1) is EPESS-active whereas Ir(ppy)(2)(L2) exhibits moderately intense emission both in solution and as a neat film, indicating that the degree of conjugation of the β-diketone ligands determines the EPESS-activity. The single-crystal X-ray analysis has indicated that there are π-π interactions between the adjacent ppy ligands in Ir(ppy)(2)(L1) but not in Ir(ppy)(2)(L2). Finally, EPESS-active Ir(ppy)(2)(L1) has been successfully embedded in polymer nanoparticles and used as a luminescent label in bioimaging.
Sigma-Aldrich
2-Phenylpyridine, 98%