Merck
  • Calcium channel subtypes for cholinergic and nonadrenergic noncholinergic neurotransmission in isolated guinea pig trachea.
Calcium channel subtypes for cholinergic and nonadrenergic noncholinergic neurotransmission in isolated guinea pig trachea.
Naunyn-Schmiedeberg's archives of pharmacology (2010-09-08)
Chung-Hung Shih, Hsin-Te Hsu, Kuo-Hsien Wang, Chih-Hsieh Shih, Wun-Chang Ko
ABSTRACT
The Ca(2+) channel subtypes in the neurotransmission of isolated guinea pig trachea were elucidated by monitoring the effects of specific Ca(2+) channel blockers on cholinergic contractions and nonadrenergic noncholinergic (NANC) relaxation elicited by electrical field stimulation (EFS). In isolated guinea pig trachea, cholinergic contractile responses to low- and high-frequency EFS were inhibited by the selective N-type calcium channel blocker, ω-conotoxin MVIIA. ω-Agatoxin IVA (a selective P-type blocker), ω-conotoxin MVIIC (a nonselective N-, Q-, and P-type blocker), and nifedipine (a selective L-type blocker) were ineffective, whereas Ni(2+) (a T- and R-type blocker) facilitated cholinergic contractions and produced a late contracture when its concentration exceeded 30 μM. The more the concentration of Ni(2+) increased, the greater the number of incidences and the late contracture areas which occurred. Late contracture did not seem to be due to the effects of acetylcholine, tachykinins, or other polypeptides, but disappeared in the absence of indomethacin. The NANC relaxant responses elicited by the low- and high-frequency EFS were inhibited by ω-conotoxin MVIIA or Ni(2+), but unaffected by ω-Agatoxin IVA, ω-conotoxin MVIIC, and nifedipine. In the absence of indomethacin, Ni(2+) did not alter the ω-conotoxin MVIIA (100 nM)-resistant component of cholinergic contraction, but significantly further inhibited that of NANC relaxation. These results suggest that in isolated guinea pig trachea, cholinergic contraction is regulated by N-type calcium channels which may mask T- and R-type calcium channels and may be co-modulated by both, while NANC relaxation is mainly and independently controlled by N-, T-, and R-type calcium channels.