Merck
  • Origin of thermodynamic versus kinetic control of allene hydroboration with 9-borabicyclo[3.3.1]nonane and 10(R)-trimethylsilyl-9-borabicyclo[3.3.2]decane.
Origin of thermodynamic versus kinetic control of allene hydroboration with 9-borabicyclo[3.3.1]nonane and 10(R)-trimethylsilyl-9-borabicyclo[3.3.2]decane.
Organic letters (2009-11-11)
Daniel H Ess, Jeremy Kister, Ming Chen, William R Roush
ABSTRACT
Density functional theory was used to locate transition states for hydroboration reactions of allenes with 9-borabicyclo[3.3.1]nonane and 10-R-9-borabicyclo[3.3.2]decane, as well as transition states for [1,3]-boratropic shift and aldehyde addition reactions of the derived allylboranes. The origin of kinetic versus thermodynamic control in the allene hydroboration step is described.
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Supelco
Nonane, analytical standard