Merck
  • Phase II study of aminothiadiazole in advanced squamous cell carcinoma of the esophagus.
Phase II study of aminothiadiazole in advanced squamous cell carcinoma of the esophagus.
American journal of clinical oncology (1991-02-01)
P F Engstrom, L M Ryan, G Falkson, D G Haller
ABSTRACT
Twenty-three patients with advanced inoperable squamous cell carcinoma of the esophagus were treated with aminothiadiazole (A-TD) 125 mg/m2 weekly plus allopurinol daily in a phase II cooperative group trial. No patients responded to treatment; 17 patients progressed, three showed stable disease, and three were unevaluable. There were no life-threatening hematologic or metabolic toxicities. The median survival from study entry was 5 months. A-TD is not active in advanced squamous cell carcinoma of the esophagus.
Sigma-Aldrich
2-Amino-1,3,4-thiadiazole, 97%