Merck
  • Preparation of partially benzylated mono-, di-, and trisaccharides by selective cleavage of the beta-fructofuranosidic linkage in fully benzylated sucrose and sucrose-related oligosaccharides under acidic conditions.
Preparation of partially benzylated mono-, di-, and trisaccharides by selective cleavage of the beta-fructofuranosidic linkage in fully benzylated sucrose and sucrose-related oligosaccharides under acidic conditions.
Carbohydrate research (2008-04-22)
Takashi Yamanoi, Noriko Misawa, Sho Matsuda, Mikio Watanabe
ABSTRACT
Several partially benzylated mono-, di-, and trisaccharides having an anomeric hydroxyl group were successfully prepared by selective cleavage of the beta-fructofuranosidic linkage in fully benzylated sucrose and sucrose-related oligosaccharides derived from lactosucrose, raffinose, melezitose, stachyose, and nystose under acidic conditions using 1:10 75% aqueous sulfuric acid-dioxane at room temperature for 1h.
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%