Merck
  • Estimating gluconeogenesis by NMR isotopomer distribution analysis of [13C]bicarbonate and [1-13C]lactate.
Estimating gluconeogenesis by NMR isotopomer distribution analysis of [13C]bicarbonate and [1-13C]lactate.
NMR in biomedicine (2007-08-09)
Tiago Cardoso Alves, Patrícia Maria Nunes, Carlos Marques Palmeira, John Griffith Jones, Rui Albuquerque Carvalho
ABSTRACT
The gluconeogenic contribution to glucose production in livers isolated from rats fasted for 24 h was determined by 13C-NMR isotopomer distribution analysis of secreted glucose enriched from 99% [13C]bicarbonate (n = 4) and 99% [1-13C]lactate (n = 4). Experiments with 3% 2H2O were also performed, allowing the gluconeogenic contribution to be measured by the relative 2H enrichments at positions 5 and 2 of glucose. From 13C-NMR analyses, the contribution of gluconeogenesis to glucose output was estimated to be 93 +/- 3% for [13C]bicarbonate perfusion and 91 +/- 3% for [1-13C]lactate perfusion, in good agreement with the 2H-NMR analysis of the gluconeogenic contribution to glucose production (100 +/- 1% and 99 +/- 1%, respectively) and consistent with the expected negligible contribution from glycogenolysis. These results indicate that 13C-NMR analysis of glucose 13C-isotopomer distribution from either [13C]bicarbonate or [1-13C]lactate precursor provides realistic estimates of the gluconeogenic contribution to hepatic glucose output.