Merck
  • "Green odor" inhalation reduces the skin-barrier disruption induced by chronic restraint stress in rats: physiological and histological examinations.
"Green odor" inhalation reduces the skin-barrier disruption induced by chronic restraint stress in rats: physiological and histological examinations.
Chemical senses (2007-06-15)
Mika Fukada, Toshiyuki Kaidoh, Ai Ito, Tomomi Yano, Chie Hayashibara, Tatsuo Watanabe
ABSTRACT
We investigated whether inhalation of green odor (a mixture of equal amounts of trans-2-hexenal and cis-3-hexenol) prevents the skin-barrier disruption induced by chronic restraint stress in rats. To this end, transepidermal water loss (TEWL) was measured as an index of the disruption of skin-barrier function, whereas light- and electron-microscope examinations were performed to observe histological changes in the skin of the stressed animals. In addition, the effects on TEWL induced by chronic administration of a glucocorticoid, dexamethasone (DEX), were examined. Chronic restraint stress (8 h per day for 14 days) increased TEWL (vehicle + stress group). This effect (and the chronic stress-induced increase in adrenal weight) was prevented in rats that inhaled green odor at the beginning of each day's restraint (2 h each day for 14 days; green odor + stress group). Electron-microscope studies revealed that rats in the green odor + stress group possessed sufficient intercorneocyte lipids to create an effective skin barrier, although these had apparently been decreased in the vehicle + stress group. Daily administration of DEX for 14 days increased TEWL. The present results suggest that chronic stress-induced disruption of the skin barrier in rats can be reduced or prevented by green odor (possibly at least in part through an inhibitory effect on the stress-induced activation of the hypothalamo-pituitary-adrenocortical axis).
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Supelco
cis-3-Hexen-1-ol, analytical standard
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%