Merck
  • [Determination of dehydroacetic acid and its sodium salt in food products and polymeric packing materials].
[Determination of dehydroacetic acid and its sodium salt in food products and polymeric packing materials].
Voprosy pitaniia (2007-02-23)
K I Eller, V V Pimenova, S V Volkovich, M G Kiseleva, A G Snezhko, Z S Borisova, V N Novikova
ABSTRACT
HPLC method of determination of dehydroacetic acid and its sodium salt in food products and polymeric packing materials has been developed. The migration of DGA from food-contacting materials to food matrix has been investigated.
Sigma-Aldrich
Dehydroacetic acid, ≥98.0% (T)