Merck
  • Solubilization of zinc compounds by the diazotrophic, plant growth promoting bacterium Gluconacetobacter diazotrophicus.
Solubilization of zinc compounds by the diazotrophic, plant growth promoting bacterium Gluconacetobacter diazotrophicus.
Chemosphere (2006-09-08)
V S Saravanan, M Madhaiyan, M Thangaraju
ABSTRACT
Gluconacetobacter diazotrophicus an endophytic diazotroph also encountered as rhizosphere bacterium is reported to possess different plant growth promoting characteristics. In this study, we assessed the zinc solubilizing potential of G. diazotrophicus under in vitro conditions with different Zn compounds using glucose or sucrose as carbon sources. G. diazotrophicus showed variations in their solubilization potential with the strains used and the Zn compounds tested. G. diazotrophicus PAl5 efficiently solubilized the Zn compounds tested and ZnO was effectively solubilized than ZnCO(3) or Zn(3)(PO(4))(2). The soluble Zn concentration was determined in the culture supernatant through Atomic Absorption Spectrophotometer. Gas chromatography coupled Mass Spectrometry analysis revealed 5-ketogluconic acid, a derivative of gluconic acid as the major organic acid produced by G. diazotrophicus PAl5 cultured with glucose as carbon source. This organic anion may be an important agent that helped in the solubilization of insoluble Zn compounds.
Sigma-Aldrich
5-Keto-D-gluconic acid potassium salt, ≥98.0% (TLC)