Merck
  • Spectroscopic and thermal studies of chromium(III), molybdenum(VI) and ruthenium(0) complexes of maleic hydrazide.
Spectroscopic and thermal studies of chromium(III), molybdenum(VI) and ruthenium(0) complexes of maleic hydrazide.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-02-07)
Hassan A Mohamed, Saadia A Ali, Ramadan M Ramadan
ABSTRACT
Interaction of maleic hydrazide (LH(2)) with [Cr(CO)(6)] in air at atmospheric pressure resulted in the formation of the complex [(LH)Cr(mu-O)(2)Cr(LH)] (1). Reaction of LH(2) with [Mo(CO)(6)] in air also gave the complex [(LH(2))O(2)Mo(mu-O)(2)MoO(2)(LH(2))] (2). Under the same conditions, the reaction of LH(2) with [Ru(3)(CO)(12)] resulted in the formation of the tricarbonyl complex [Ru(CO)(3)(LH(2))] (3). The complexes were characterized by elemental analysis, IR, and (1)H NMR spectroscopy. The thermal properties of the complexes were investigated by thermogravimetry technique.
Sigma-Aldrich
Maleic hydrazide, ReagentPlus®, 99%