Merck
Effects of mushroom tyrosinase on anisaldehyde.
Journal of agricultural and food chemistry (2005-09-01)
Tae Joung Ha, Sadaaki Tamura, Isao Kubo
ABSTRACT
Anisaldehyde (p-methoxybenzaldehyde) was previously reported to inhibit the tyrosinase-catalyzed oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) noncompetitively as long as the enzyme activity was monitored by measuring dopachrome formation. However, anisaldehyde did not inhibit this oxidation if a longer reaction time was observed, although it suppressed the initial rate of oxidation to a certain extent. Anisaldehyde significantly suppressed the rate of enzymatic oxidation of L-tyrosine.
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
p-Anisaldehyde, 98%
Supelco
p-Anisaldehyde, analytical standard