Merck
  • Rapid determination of C6-aldehydes in tomato plant emission by gas chromatography-mass spectrometry and solid-phase microextraction with on-fiber derivatization.
Rapid determination of C6-aldehydes in tomato plant emission by gas chromatography-mass spectrometry and solid-phase microextraction with on-fiber derivatization.
Journal of separation science (2005-03-10)
Chunhui Deng, Ning Li, Weimin Zhu, Ji Qian, Xiaofeng Yang, Xiangmin Zhang
ABSTRACT
A simple, rapid, sensitive, and solvent-free method was developed for determination of plant-signalling compounds, the three C6-aldehydes hexanal, (Z)-3-hexenal, and (E)-2-hexenal, in tomato plant emission by gas chromatography-mass spectrometry (GC-MS) and solid-phase microextraction (SPME) with on-fiber derivatization. In this method, O-2,3,4,5,6-(pentafluorobenzyl)hydroxylamine (PFBHA) in aqueous solution was first headspace adsorbed onto a 65 microm poly(dimethylsiloxane)/divinylbenzene (PDMS/DVB) fiber at 25 degrees C for 5 min, and then the fiber with adsorbed PFBHA was used for headspace extraction of tomato plant emission at 25 degrees C for 6 min. Finally, the resulting oximes adsorbed on the fiber were desorbed and analyzed by GC-MS. Extraction conditions and method validation were studied. The proposed method had low detection limit values for the three aldehydes from 0.1 to 0.5 ng/L and good precision (RSD less than 10%). In this work, the method was applied to investigation of tomato plant defense response to Helicoverpa armigera.
Product Description
Supelco
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, derivatization grade (GC derivatization), LiChropur, ≥99.0% (AT)
Sigma-Aldrich
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, ≥98%