Merck
Solvents and vapor intrusion pathways.
Clinics in occupational and environmental medicine (2004-08-25)
Scott D Phillips, Gary R Krieger, Robert B Palmer, Javier C Waksman
ABSTRACT
Vapor intrusion must be recognized appropriately as a separate pathway of contamination. Although many issues resemble those of other forms of contamination (particularly its entryway, which is similar to that of radon seepage), vapor intrusion stands apart as a unique risk requiring case-specific action. This article addresses these issues and the current understanding of the most appropriate and successful remedial actions.
Product Description
Supelco
1,1-Dichloroethylene, analytical standard